missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMBR1 Monoclonal Antibody (4A1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA522489
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: SRMFTVMGQL LVKPTILEDL DEQIYIITLE EEALQRRLNG LSSSVEYNIM ELEQELENVK TLKTKLERRK KASAWERNLV YP
LMBR1 Monoclonal antibody specifically detects LMBR1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
LMBR1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8WVP7 | |
Lmbr1 | |
LMBR1 (NP_071903, 214 a.a. ∽ 295 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4A1 | |
Unconjugated | |
Lmbr1 | |
1110048D14Rik; ACHP; AU017641; C7orf2; DIF14; Differentiation-related gene 14 protein; FLJ11665; limb development membrane protein 1; limb region 1; limb region 1 homolog; limb region 1 protein; Limb region 1 protein homolog; Lmbr1; LSS; PPD2; THYP; TPT; Unknown (protein for MGC:134350); ZRS | |
Mouse | |
Protein A | |
RUO | |
64327 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |