Learn More
LOX-1 Monoclonal Antibody (331212), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524060
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse LOX-1. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
LOX-1 Monoclonal antibody specifically detects LOX-1 in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
LOX-1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P78380 | |
OLR1 | |
Mouse myeloma cell line NS0-derived recombinant human LOX-1/OLR1 Ser61-Gln273 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
331212 | |
Unconjugated | |
OLR1 | |
CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLOX1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LOX1; LOX-1; LOXIN; Oldlr1; Oldr1; OLR1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; Scare1; scavenger receptor class E, member 1; sLOX 1; SLOX1; soluble LOX 1; SR-EI | |
Mouse | |
Protein A/G | |
RUO | |
4973 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |