missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOXL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532900
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Marmoset (100%) Gorilla, Orangutan, Monkey, Bovine, Bat, Elephant (94%) Dog, Panda, Horse, Opossum (88%).
LOXL2 Polyclonal antibody specifically detects LOXL2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
LOXL2 | |
Polyclonal | |
Unconjugated | |
LOXL2 | |
1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9Y4K0 | |
LOXL2 | |
Synthetic 17 amino acid peptide from internal region of human LOXL2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |