missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPCAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539008
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes a member of the lysophospholipid acyltransferase family. The encoded enzyme may function in two ways: to catalyze the biosynthesis of platelet-activating factor (1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) from 1-O-alkyl-sn-glycero-3-phosphocholine, and to catalyze the synthesis of glycerophospholipid precursors from arachidonyl-CoA and lysophosphatidylcholine. The encoded protein may function in membrane biogenesis and production of platelet-activating factor in inflammatory cells. The enzyme may localize to the endoplasmic reticulum and the Golgi. [provided by RefSeq, Feb 2009].Spezifikation
LPCAT2 | |
Polyclonal | |
Unconjugated | |
LPCAT2 | |
Aytl1, Aytl1a, Lpcat2a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100359680, 270084, 54947 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7L5N7, Q8BYI6, P0C1Q3 | |
LPCAT2 | |
A synthetic peptide derived from the internal region of human LPCAT2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |