missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPP Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519230
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and canine based on sequence homology.
LPP Polyclonal antibody specifically detects LPP in Human samples. It is validated for Western Blot.Spezifikation
LPP | |
Polyclonal | |
Unconjugated | |
LPP | |
9430020K16Rik; AA959454; AU024130; B130055L10Rik; C79715; D630048H16; I79_000864; LIM domain containing preferred translocation partner in lipoma; LIM domain-containing preferred translocation partner in lipoma; LIM protein; lipoma preferred partner; Lipoma-preferred partner; Lipoma-preferred partner homolog; Lipoma-preferred-partner; LPP | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4026 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q93052 | |
LPP | |
Synthetic peptide sequence (RNDSDPTYGQQGHP) corresponding to the internal amino acids of LPP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |