Learn More
Lano Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531722
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116, 293T, A431. Predicted reactivity: Mouse (87%), Rat (87%), Rhesus Monkey (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lano Polyclonal antibody specifically detects Lano in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Lano | |
Polyclonal | |
Unconjugated | |
LRRC1 | |
A430093J20Rik; AU016091; D030059I21; dJ523E19.1; LANO; LANO adapter protein; LAP (leucine-rich repeats and PDZ) and no PDZ protein; LAP and no PDZ protein; leucine rich repeat containing 1; leucine-rich repeat-containing 1; leucine-rich repeat-containing protein 1; LRRC1; mKIAA4018; RP3-523E19.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BTT6 | |
LRRC1 | |
Recombinant fragment corresponding to a region within amino acids 265 and 524 of Human LRRC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |