missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRFIP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521115
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is rat colon lysate. PA5-21115 can be used with blocking peptide PEP-1229.
LRRFIP2 Polyclonal antibody specifically detects LRRFIP2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
LRRFIP2 | |
Polyclonal | |
Unconjugated | |
LRRFIP2 | |
5133400F20Rik; AI850587; HUFI-2; leucine rich repeat (in FLII) interacting protein 2; leucine-rich repeat flightless-interactin protein 2-like protein; leucine-rich repeat flightless-interacting protein 2; LRR binding FLII interacting protein 2; LRR FLII-interacting protein 2; LRRFIP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
301035, 71268, 9209 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q4V7E8, Q91WK0, Q9Y608 | |
LRRFIP2 | |
A 17 amino acid peptide near the carboxy terminus of human LRRFIP2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |