missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA513868
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
LRRK1 Polyclonal antibody specifically detects LRRK1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
LRRK1 | |
Polyclonal | |
Unconjugated | |
Lrrk1 | |
AW319595; C230002E15Rik; D130026O16Rik; Kiaa1790; leucine rich repeat kinase 1; leucine-rich repeat kinase 1; leucine-rich repeat serine/threonine-protein kinase 1; LRRK1; mKIAA1790; RIPK6; Roco1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
233328, 79705 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q38SD2, Q3UHC2 | |
Lrrk1 | |
KLH conjugated synthetic peptide between 1981-2015 amino acids from the C-terminal region of human LRRK1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |