Learn More
LSM4, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA128314
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
PA1-28314 detects LSM4 from mouse, human, rat samples.PA1-28314 has been successfully used in Western blot applications.The PA1-28314 immunogen is: Recombinant fragment, corresponding to amino acids 1-139 of Human LSM4.Spezifikation
LSM4 | |
Polyclonal | |
Unconjugated | |
Q9Y4Z0, Q9QXA5 | |
LSM4 | |
Recombinant fragment, corresponding to amino acids 1-139 of Human LSM4. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
ELISA, Western Blot | |
1mg/mL | |
LSM4 | |
YER112W | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
25804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |