Learn More
LTBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545075
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NVCANGDCSN LEGSYMCSCH KGYTRTPDHK HCRDIDECQQ GNLCVNGQCK Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 83%; Horse: 86%; Human: 100%; Mouse: 83%; Rabbit: 92%; Rat: 83%
LTBP1 Polyclonal antibody specifically detects LTBP1 in Human samples. It is validated for Western Blot.Spezifikation
LTBP1 | |
Polyclonal | |
Unconjugated | |
LTBP1 | |
9430031G15Rik; 9830146M04; b2b1000Clo; LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; latent transforming growth factor beta binding protein 1; latent transforming growth factor binding protein 1; latent transforming growth factor binding protein-1; latent-transforming growth factor beta-binding protein 1; LTBP1; LTBP-1; MGC163161; Tgfb; TGF-beta-1-BP-1; TGF-beta1-BP-1; transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4052 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14766 | |
LTBP1 | |
Synthetic peptide directed towards the C-terminal of human LTBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |