Learn More
Firefly luciferase Monoclonal Antibody (LUC-Y), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA180225
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Membrane permeabilization is required for flow cytometry applications. In Western Blot , MA1-80225 detects a band at ∽55-60 kDa using 1 μg of purified luciferase. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Luciferase antibody, clone LUC-Y detects the expression of the luciferase gene of the North American firefly Photinus pyralis in cells and tissues.
Firefly luciferase Monoclonal antibody specifically detects Firefly luciferase in Luciferase samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Firefly luciferase | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Luciferase | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
LUC-Y | |
Unconjugated | |
Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase | |
Recombinant luciferase expressed in E. Coli | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |