missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LUM Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547410
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Lumican is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
LUM Polyclonal antibody specifically detects LUM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
LUM | |
Polyclonal | |
Unconjugated | |
LUM | |
keratan sulfate proteoglycan lumican; KSPG lumican; Lcn; Ldc; Lum; Lumican; Lumican precursor; lumican proteoglycan; SLRR2D | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4060 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P51884 | |
LUM | |
Mouse myeloma cell line NS0-derived recombinant human Lumican Gln19-Asn338 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |