Learn More
Luteinizing Hormone-beta Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Marke: Invitrogen PA118415
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Reconstitute in 50 μL sterile water. Centrifuge to remove any insoluble material. It is recommended that a thawed sample is stored at 4 °C for no longer than 2 weeks. Allocation of appropriate anti-bacterial agent can increase shelf life by several weeks. Diluted serum should be prepared as required. Long term stability requires storage, preferably in small aliquots at -20 °C or lower. Glycerol (1:1) can be added to neat serum for additional stability if intended use does not prevent this.
Luteinizing Hormone-beta Polyclonal antibody specifically detects Luteinizing Hormone-beta in Ovine samples. It is validated for Immunohistochemistry (Frozen).Spezifikation
Luteinizing Hormone-beta | |
Polyclonal | |
Unconjugated | |
LHB | |
CGB4, LSH-B, hLHB | |
Guinea Pig | |
50 μL | |
Primary | |
Ovine | |
Antibody | |
IgG |
Immunohistochemistry (Frozen) | |
Conc. Not Determined | |
whole serum with no preservative | |
P01231 | |
LHB | |
Synthetic peptide corresponding to the antigenic region within the ovine LH residue. | |
RUO | |
443395 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |