missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteinizing Hormone-beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA123783
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Human anterior pituitary. Previous lots of this antibody have been shown to react with rat and monkey samples but this has not been tested.
Luteinizing Hormone-beta Polyclonal antibody specifically detects Luteinizing Hormone-beta in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry.Spezifikation
Luteinizing Hormone-beta | |
Polyclonal | |
Unconjugated | |
LHB | |
LH | |
Rabbit | |
Protein A | |
RUO | |
3972 | |
4° C, do not freeze | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
2.0 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide | |
P01229 | |
LHB | |
Human luteinizing hormone(LH) | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |