Learn More
anti-LXR alpha/NR1H3 Ligand Binding Domain, Clone: PPZ0412, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) PP-PPZ0412-00
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412476
  • 588.77 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Applications:
Gel Supershift, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
LXR alpha/NR1H3
PPZ0412
Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Supershift Assay
Purified
NR1H3
IgG2a
100 ug
Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Monoclonal
Human
Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Shift
Unconjugated
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, NR1H3, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10062
Videos