Learn More
Lymphotactin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA184207
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. Reconstitute with 0.1 mL of distilled water. For long term storage the addition of 0.09% sodium azide is recommended. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. Rabbit anti Human Lymphotactin antibody recognizes human Lymphotactin, otherwise known as XCL1, the only member of the C-chemokine family identified so far.
Lymphotactin Polyclonal antibody specifically detects Lymphotactin in Human samples. It is validated for ELISA, Functional Assay, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Lymphotactin | |
Polyclonal | |
Unconjugated | |
Xcl1 | |
AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM1alpha; SCM-1alpha; SCM-1-alpha; SCM1beta; SCM-1beta; SCM-1-beta; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6375 | |
-20°C | |
Lyophilized |
ELISA, Functional Assay, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P47992 | |
Xcl1 | |
Recombinant human lymphotatin | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |