Learn More
Lyn Monoclonal Antibody (2H8D7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515329
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15329 targets LYN in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15329 immunogen is purified recombinant fragment of LYN expressed in E. Coli. MA5-15329 detects LYN which has a predicted molecular weight of approximately 56kDa.
Lyn Monoclonal antibody specifically detects Lyn in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Lyn | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P07948 | |
LYN | |
Purified recombinant fragment of LYN expressed in E. Coli. | |
RUO | |
4067 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
2H8D7 | |
Unconjugated | |
LYN | |
AA407514; CD180; CD180 antigen; CD180 molecule; CH73-38P6.3; FLJ26625; Hck-2; JTK8; lck/Yes-related novel protein tyrosine kinase; LY64; Ly78; lymphocyte antigen 64; lymphocyte antigen-64, radioprotective, 105kDa; LYN; lyn protein non-receptor kinase; lyn protein tyrosine kinase; LYN proto-oncogene, Src family tyrosine kinase; p53Lyn; p56Lyn; Radioprotective 105 kDa protein; RP105; Tyrosine-protein kinase Lyn; v-yes-1 Yamaguchi sarcoma viral related oncogene homolog; v-yes-1 Yamaguchi sarcoma viral related oncogene protein-like protein; Yamaguchi sarcoma viral (v-yes-1) oncogene homolog; zgc:92124 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |