missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532908
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Dog, Elephant, Horse, Rabbit (100%) Marmoset, Mouse, Rat, Panda, Pig (93%) Bovine (86%).
LYPD3 Polyclonal antibody specifically detects LYPD3 in Canine, Human, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
LYPD3 | |
Polyclonal | |
Unconjugated | |
LYPD3 | |
2310061G07Rik; C4.4A; GPI-anchored metastasis-associated protein C4.4A homolog; GPI-anchored metastasis-associated protein homolog; LY6/PLAUR domain containing 3; ly6/PLAUR domain-containing protein 3; LYPD3; Matrigel-induced gene C4 protein; MIG-C4; UNQ491/PRO1007 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100339878, 27076, 484471 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O95274 | |
LYPD3 | |
Synthetic 14 amino acid peptide from N-terminus of human LYPD3. | |
50 μg | |
Primary | |
Canine, Human, Rabbit | |
Antibody | |
IgG |