Learn More
Lysozyme Monoclonal Antibody (BGN/0696/5B1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA182873
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. A suggested positive control for this product is human tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Lysozyme antibody, clone BGN/0696/5B1 recognizes lysozyme, an enzyme that is present in mucosal secretions such as saliva and tears and is involved with non-specific organism defence.
Lysozyme Monoclonal antibody specifically detects Lysozyme in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
Lysozyme | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P61626 | |
LYZ | |
Native protein, from urine. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
BGN/0696/5B1 | |
Unconjugated | |
LYZ | |
1,4-beta-N-acetylmuramidase C; 1700038F02Rik; 4-beta-N-acetylmuramidase C; AI326280; Allergen Gal d IV; bA534G20.1; c-type lysozyme; dystrophin; egg white lysozyme; Gal d 4; KAAG648; LYC2; Lys; Lysm; lysozyme; lysozyme (renal amyloidosis); lysozyme 1; lysozyme 2; lysozyme C; Lysozyme C type M; lysozyme C type P; Lysozyme C, spleen isozyme; lysozyme C-1; Lysozyme C-2; lysozyme C-3; lysozyme d1; lysozyme F1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; Lysz; LYZ; Lyz1; Lyz2; LYZC; LYZD1; LYZF1; LYZL1; Lyzs; Lzm; Lzm-s1; Lzp; Lzp-s; P lysozyme structural; PRO1278; renal amyloidosis; RGD1559869; unnamed protein product; UNQ648/PRO1278 | |
Mouse | |
Protein A | |
RUO | |
4069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |