Learn More
MAdCAM-1 Rat anti-Mouse, Clone: 200530, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB9931
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412086
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
MAdCAM-1 Monoclonal antibody MAB9931 Specifically detects MAdCAM-1 in Mouse samples. It is validated for ELISA Capture
Spezifikation
Spezifikation
MAdCAM-1
200530
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
NP_038619
hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse MAdCAM-1 in ELISAs.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MADCAM1
Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365 (predicted) Accession # NP_038619
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8174
Reconstitute at 0.5 mg/mL in sterile PBS.