missing translation for 'onlineSavingsMsg'
Learn More

FABP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532487

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13427007

  • 339.00 € / 500 Mikroliter


Voraussichtlicher Liefertermin 06-09-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

MDGI /FABP3 is a small cytosolic protein responsible for the transport and deposition of fatty acids inside the cell.
Spezifikation

Spezifikation

FABP3
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P05413
FABP11, H-FABP, M-FABP, MDGI, O-FABP, Fatty acid-binding protein 3, muscle, fatty acid binding protein 11, fatty acid-binding protein, heart, heart-type fatty acid-binding protein, mammary-derived growth inhibitor, muscle fatty acid-binding protein
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
2170
Immunohistochemistry (Paraffin)
Unconjugated
FABP3
Liquid
FABP3
Synthetic peptide derived from N-terminus of human MDGI protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Dokumentation