Learn More
MOCOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530662
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, HepG2. Predicted reactivity: Mouse (83%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MOCOS Polyclonal antibody specifically detects MOCOS in Human samples. It is validated for Western Blot.Spezifikation
MOCOS | |
Polyclonal | |
Unconjugated | |
MOCOS | |
1110018O12Rik; HMCS; MCS; moCo sulfurase; MOCOS; Molybdenum cofactor sulfurase; molybdenum cofactor sulfurtransferase; MOS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96EN8 | |
MOCOS | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 283 of Human MOCOS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |