Learn More
MOCS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531366
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Pig (86%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MOCS3 Polyclonal antibody specifically detects MOCS3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MOCS3 | |
Polyclonal | |
Unconjugated | |
MOCS3 | |
1700020H17Rik; Adenylyltransferase and sulfurtransferase MOCS3; Adenylyltransferase MOCS3; dJ914P20.3; MOCS3; molybdenum cofactor synthesis 3; molybdenum cofactor synthesis protein 3; Molybdopterin synthase sulfurylase; Molybdopterin-synthase adenylyltransferase; Molybdopterin-synthase sulfurtransferase; MPT synthase sulfurylase; Sulfur carrier protein MOCS2A adenylyltransferase; Sulfur carrier protein MOCS2A sulfurtransferase; Sulfurtransferase MOCS3; Uba4; UBA4, ubiquitin-activating enzyme E1 homolog; ubiquitin-like modifier activating enzyme 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27304 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O95396 | |
MOCS3 | |
Recombinant fragment corresponding to a region within amino acids 220 and 460 of Human MOCS3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |