missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOGAT2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519372
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
MOGAT2 Polyclonal antibody specifically detects MOGAT2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
MOGAT2 | |
Polyclonal | |
Unconjugated | |
MOGAT2 | |
2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2; acyl-CoA:monoacylglycerol acyltransferase 2; DC5; Dgat2l5; DGAT2L5.; Diacylglycerol acyltransferase 2-like protein 5; diacylglycerol O-acyltransferase candidate 5; FLJ22644; hDC5; hMGAT2; Mgat1l; MGAT2; MGC119183; MGC119184; MGC119185; MOGAT2; Monoacylglycerol O-acyltransferase 1-like; Monoacylglycerol O-acyltransferase 2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80168 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q3SYC2 | |
MOGAT2 | |
Synthetic peptide sequence (KESAAHILNRK) corresponding to the internal amino acids of MOGAT2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |