Learn More
Macrophages/Monocytes Monoclonal Antibody (ER-HR3), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA140078
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-40078 detects Monocytes/Macrophages from mouse samples. MA1-40078 is specially reactive to the monocytes and macrophages located in hemopoietic organs. Whereas all other leukocytes are ER-HR3 negative, up to 70% of circulating monocytes stain positive with this monoclonal antibody. Furthermore, about 30% of bone marrow cells express the antigen. Other positive macrophages can be found in the splenic red pulp, in the mesenteric lymphoid paracortex, interfollicular areas of Peyer’s patches and bone marrow. Few positive cells are observed in the thymic cortex and the connective tissues of the gastro-intestinal tract, the dermis and the renal medulla. Epidermal Langerhans cells also express the antigen. MA1-40078 has been successfully used in immunohistochemistry (paraffin and frozen) and flow cytometry applications. The MA1-40078 immunogen is mouse monocytes and macrophages.
Macrophages/Monocytes Monoclonal antibody specifically detects Macrophages/Monocytes in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
Macrophages/Monocytes | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
Rat | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG2c |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
ER-HR3 | |
Unconjugated | |
Macrophage; Monocyte | |
Murine monocytes and macrophages. | |
100 μg | |
Primary | |
4° C | |
Liquid |