Learn More
Activin RIB/ALK-4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1477
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416964
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Activin RIB/ALK-4 Polyclonal antibody AF1477 Specifically detects Activin RIB/ALK-4 in Mouse samples. It is validated for Blocking (Receptor-ligand), Immunohistochemistry, Western Blotting
Spezifikation
Spezifikation
Activin RIB/ALK-4
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
Q61271
activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Activin RIB/ALK-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Activin RIB is observed, 5% cross-reactivity with rhActivin R1A is observed and less than 1% cross-reactivity with rhActivin RIIA and rhActivin RIIB is observed.
Mouse
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ACVR1B
Mouse myeloma cell line NS0-derived recombinant mouse Activin RIB/ALK-4, Leu32-Glu126, Accession # Q61271
Immunogen affinity purified
RUO
Primary
91
Reconstitute at 0.2 mg/mL in sterile PBS.