Learn More
Aminoacylase/ACY1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2700
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417734
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Aminoacylase/ACY1 Polyclonal antibody AF2700 Specifically detects Aminoacylase/ACY1 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
Aminoacylase/ACY1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
NP_079647
ACY1, ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Aminoacylase/ACY1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human Aminoacylase/ACY1 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ACY1
Mouse myeloma cell line NS0-derived recombinant mouse Aminoacylase/ACY1 Met1-Ser408 Accession # NP_079647
Antigen Affinity-purified
RUO
Primary
95
Reconstitute at 0.2 mg/mL in sterile PBS.