Learn More
Cathepsin F Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2544
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417634
  • 341.89 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Cathepsin F Polyclonal antibody AF2544 Specifically detects Cathepsin F in Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
Cathepsin F
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Q9R013
cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Cathepsin F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Cathepsin C, rmCathepsin H, and rmCathepsin L is observed.
Mouse
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CTSF
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin F, Ser20-Asn462, Accession # Q9R013
Antigen Affinity-purified
RUO
Primary
8722
Reconstitute at 0.2 mg/mL in sterile PBS.