Learn More
CD2F-10/SLAMF9 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2829
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417814
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD2F-10/SLAMF9 Polyclonal antibody AF2829 Specifically detects CD2F-10/SLAMF9 in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
CD2F-10/SLAMF9
LYOPH
Western Blot 0.1 ug/mL
Q9D780
CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9, SLAMF9
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse CD2F-10/SLAMF9 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse CD2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SLAMF9
Mouse myeloma cell line NS0-derived recombinant mouse CD2F-10/SLAMF9, Phe18-Leu230, Accession # Q9D780
Antigen Affinity-purified
RUO
Primary
89886
Reconstitute at 0.2 mg/mL in sterile PBS.