Learn More
CD59b Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2650
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417704
  • 491.42 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD59b Polyclonal antibody AF2650 Specifically detects CD59b in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
CD59b
LYOPH
Western Blot 0.1 ug/mL
P58019
Cd59b
Mouse myeloma cell line NS0-derived recombinant mouse CD59b Leu24-Asn104 Accession # P58019
Immunogen affinity purified
RUO
Primary
966
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse CD59b in direct ELISAs and Western blots.