Learn More
HRP-1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1868
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417204
  • 324.73 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
HRP-1 Polyclonal antibody AF1868 Specifically detects HRP-1 in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
HRP-1/HDGFL1
LYOPH
Western Blot 0.1 ug/mL
NP_032258
dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, HRP1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse HRP-1 in direct ELISAs and Western blots.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HDGFL1
E. coli-derived recombinant human HRP-1 Ser2-Leu283 Accession # NP_032258
Antigen Affinity-purified
RUO
Primary
154150
Reconstitute at 0.2 mg/mL in sterile PBS.