Learn More
Iduronate 2-Sulfatase/IDS Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2486
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417594
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Iduronate 2-Sulfatase/IDS Polyclonal antibody AF2486 Specifically detects Iduronate 2-Sulfatase/IDS in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
Iduronate 2-Sulfatase/IDS
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q08890
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Iduronate 2-Sulfatase/IDS in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human IDS is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IDS
Mouse myeloma cell line NS0-derived recombinant mouse Iduronate 2-Sulfatase/IDS, Thr36-Pro552, Accession # Q08890
Antigen Affinity-purified
RUO
Primary
3423
Reconstitute at 0.2 mg/mL in sterile PBS.