missing translation for 'onlineSavingsMsg'
Learn More

F(ab')2-Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, AP, Invitrogen™

F(ab')2-Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen PA184469

378.68 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11599160

  • 568.00 € / 500 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Cross reactivity is observed with rat IgG. Cross-reactivity with human IgG has been minimized by solid phase absorption. F(ab')2 fragments were prepared by pepsin digestion of the IgG followed by a gel filtration step to remove the remaining intact IgG or Fc fragments. Rabbit anti Mouse IgG antibody recognizes mouse IgG.Cross reactivity is observed with rat IgG. Cross-reactivity with human IgG has been minimised by solid phase absorption.

Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
Alkaline Phosphatase (AP)
Rabbit
Affinity chromatography
RUO
Mouse
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 1% BSA, 25% glycerol and <0.1% sodium azide; pH 8.0
Murine IgG.
500 μg
Secondary
4° C, do not freeze
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote