Learn More
Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 350, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen A21062
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
Product will be shipped at Room Temperature.
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 350 | |
Rabbit | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |