Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen G21060
Weitere Details : Netto-Gewicht : 0.00525kg
Beschreibung
The specific activity of the AP conjugates is approximately 150–350 units/mg. One unit is defined as the amount of enzymatic activity required to hydrolyze 1.0 micromole of p nitrophenyl phosphate to p-nitrophenol and inorganic phosphate in 60 seconds at 21oC, pH 10. Reconstituted using 0.5 mL of deionized water to make a 2 mg/mL stock solution.
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
purified | |
RUO | |
Mouse | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
2 mg/mL | |
50mM tris with 0.1M NaCl, 30mM trehalose and no preservative; pH 8.0 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
-20°C | |
IgG |