missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen A24528
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Biotin | |
Goat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |