missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Donkey anti-Mouse IgG (H+L) Secondary Antibody, FITC, Invitrogen™
F(ab')2-Donkey Polyclonal Secondary Antibody
Marke: Invitrogen A24501
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
Mouse IgG (H+L) | |
Polyclonal | |
FITC | |
Donkey | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |