Learn More
Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen A16171
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin mouse serum proteins, human serum proteins.
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Immunocytochemistry.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
TRITC | |
Rabbit | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |
Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |