missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Isotype Control (NCG01), Invitrogen™
Mouse Isotype Control
Marke: Invitrogen MA514453
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-14453 reacts in ELISA, flow cytometry, gel shift, immunofluorescence, immunohistochemistry (paraffin and frozen), immunoprecipitation, and western blot analysis.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Mouse IgG1 | |
NCG01 | |
Unconjugated | |
P01868 | |
Ighg1 | |
Synthetic hapten which is normally not present in humans or animals | |
500 μL | |
Isotype Control (Primary) | |
4° C | |
IgG1 κ |
Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
0.1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Mouse | |
Protein G | |
RUO | |
Not Applicable | |
Liquid |