Learn More
Mouse IgG1 Isotype Control, FITC, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen PA533202
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33202 has been tested and found to be negative on the following rat tissues - peripheral blood leucocytes, thymocytes, splenocytes and macrophages. PA5-33202 recognizes the human CD45RA antigen, and therefore human leucocytes may be used as a positive control for this product. This antibody is not suitable for use as a negative control on human tissues. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG1 | |
0.1 mg/mL | |
PBS mit 1 % BSA und 0.09 % Natriumazid | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
FITC | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Human T lymphocytes | |
100 μg | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG1 |