Learn More
Mouse IgG1 Isotype Control, PE, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen PA533203
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33203 has been tested and found to be negative on the following rat tissues - peripheral blood leucocytes, thymocytes, splenocytes and macrophages. PA5-33203 recognizes the human CD45RA antigen, and therefore human leucocytes may be used as a positive control for this product. This antibody is not suitable for use as a negative control on human tissues. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Once reconstituted, this product has a shelf life of approx. 6 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG1 | |
Conc. Not Determined | |
PBS mit 1 % BSA, 5 % Saccharose und 0.09 % Natriumazid | |
Mouse | |
Protein G | |
RUO | |
Not Applicable | |
Lyophilized |
Control, Flow Cytometry | |
PE | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Human T lymphocytes | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |