Learn More
Mouse IgG1 Isotype Control Cocktail, FITC, PE, Invitrogen™
Isotype Control Cocktail
Marke: Invitrogen PA533182
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33182 is suitable for use a negative control for measurement of non-specific binding of mouse monoclonals of isotype IgG1 in rat tissue in a dual labelling technique using FITC and R-Phycoerythrin fluorochromes. This antibody contains a monoclonal mixture of FITC-conjugated Mouse IgG1 RPE-conjugated Mouse IgG1. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG1 Cocktail antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG1 | |
Cocktail | |
FITC/PE | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Antigen not available at this time | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
Control, Flow Cytometry | |
Conc. Not Determined | |
PBS mit 1 % BSA, 1 % BSA, 5 % Saccharose, 5 % Saccharose und 0.09 % Natriumazid, 0.09 % Natriumazid | |
Mouse | |
purified | |
RUO | |
Not Applicable | |
Lyophilized |