Learn More
Mouse IgG2a Isotype Control, Invitrogen™
Mouse Isotype Control
Marke:  Invitrogen PA533208
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 15394401
  • 1156.00 € / 1 Milligramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
PA5-33208 is suitable for use as a matched isotype negative control reagent to assess the level of non-specific binding of mouse IgG2a monoclonal antibodies in rat cells. This antibody has been tested and found to be negative on the following rat cells - peripheral blood leucocytes, thymocytes, splenocytes and macrophages. PA5-33208 recognizes the human CD45 antigen and therefore human leucocytes may be used as a positive control for this product. This antibody is not suitable for use as a negative control on human tissues. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG2a Control antibody specifically detects Mouse IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry.
Spezifikation
Spezifikation
Mouse IgG2a
1 mg/mL
PBS ohne Konservierungsmittel
Mouse
Protein A
RUO
Not Applicable
Liquid
Control, Flow Cytometry
Unconjugated
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Human T lymphocytes
1 mg
Isotype Control (Primary)
-20° C, Avoid Freeze/Thaw Cycles
IgG2a