Learn More
Mouse IgG2a Isotype Control, FITC, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen PA533239
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33239 is suitable for use as a negative control reagent for the measurement of non-specific binding of mouse monoclonal antibodies of isotype IgG2a in human tissue. This antibody may not be suitable for intracellular staining on some cell types. The antibody recognizes a rat cell surface marker, and therefore cannot be used as a negative control in this species. This product is routinely tested in flow cytometry on rat splenocytes to confirm antibody activity and on human whole blood to test for suitability as a negative control. Test results have shown that PA5-33239 is also suitable for use as a negative control with bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG2a Control antibody specifically detects Mouse IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG2a | |
0.1 mg/mL | |
PBS mit 1 % BSA und 0.09 % Natriumazid | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
FITC | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Activated rat T-helper cells | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG2a |