Learn More
Mouse IgG2b Isotype Control (TEN-0), Invitrogen™
Mouse Isotype Control
Marke: Invitrogen MA190550
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-90550 does not react with any human cell surface markers, and may therefore may be used as an isotype matched negative control reagent to assess the level of non-specific binding of mouse IgG2b monoclonal antibodies in human cells. This antibody is also suitable for use as a negative control with porcine, and guinea-pig cells.
Mouse IgG2b Control antibody specifically detects Mouse IgG2b in Not Applicable samples. It is validated for Flow Cytometry.Spezifikation
Mouse IgG2b | |
TEN-0 | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Spontaneous murine plasmacytoma. | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |