Learn More
Mouse IgG2b Isotype Control, PE, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen PA533226
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33226 is a mouse IgG2b monoclonal antibody arising from a spontaneous mouse plasmacytoma. The antibody does not react with any human cell surface markers, and may therefore be used as an isotype matched negative control reagent to assess the level of non-specific binding of mouse IgG2b monoclonal antibodies in human cells. PA5-33226 is also suitable for use as a negative control with bovine, sheep, pig, canine, rat and guinea-pig tissues. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG2b Control antibody specifically detects Mouse IgG2b in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG2b | |
Conc. Not Determined | |
PBS mit 1 % BSA und 0.09 % Natriumazid | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Lyophilized |
Control, Flow Cytometry | |
PE | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Mouse IgG2b | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b |