Learn More
Mouse IgG3 Isotype Control (PPV-07), DyLight™ 650, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen MA1190D650
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
MA1-190-D650 can be used as a negative control in any immunoassay where a DyLight 650-conjugated primary antibody of the mouse IgG3 isotype is used. DyLight 650 has an excitation/emission of 652/672nm.
Mouse IgG3 Control antibody specifically detects Mouse IgG3 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Mouse IgG3 | |
PPV-07 | |
DyLight 650 | |
IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3 | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG3 |