Learn More
IL-1 Rrp2/IL-1 R6 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2354
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417504
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IL-1Rrp2/IL-1R6 Polyclonal antibody specifically detects IL-1Rrp2/IL-1R6 in Mouse samples. It is validated for Western Blot.
Spezifikation
Spezifikation
IL-1Rrp2/IL-1R6
LYOPH
Western Blot 0.1 ug/mL
Q9ERS7
Il1rl2
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 Rrp2/IL-1 R6 Asp22-Arg338 (Ala158Val) Accession # Q9ERS7
Antigen Affinity-purified
RUO
Primary
8808
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs, less than 60% with recombinant rat IL-1 Rrp2 and recombinant human IL-1 Rrp2 is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-1 R4 is observed.