Learn More
Integrin alpha 9 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3827
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418394
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Integrin alpha 9 Polyclonal antibody AF3827 Specifically detects Integrin alpha 9 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Spezifikation
Spezifikation
Integrin alpha 9
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
NP_598482
ALPHA-RLC, integrin alpha-9, Integrin alpha-RLC, integrin, alpha 9, ITGA4L, ITGA9, RLC, RLC-alpha
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Integrin alpha 9 in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human Integrin alpha 9 is observed and less than 5% cross-reactivity with recombinant mouse Integrin alpha 3 and alpha 4 is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ITGA9
Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979 Accession # NP_598482
Antigen Affinity-purified
RUO
Primary
3680
Reconstitute at 0.2 mg/mL in sterile PBS.