Learn More
Marapsin/Pancreasin Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1989
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417274
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Marapsin/Pancreasin Polyclonal antibody AF1989 Specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Spezifikation
Spezifikation
Marapsin/Pancreasin
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5 ug/mL
Q8BJR6
CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Marapsin/Pancreasin in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Marapsin is observed.
Mouse
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PRSS27
Mouse myeloma cell line NS0-derived recombinant mouse Marapsin/Pancreasin Ala23-Thr290 Accession # Q8BJR6
Immunogen affinity purified
RUO
Primary
83886
Reconstitute at 0.2 mg/mL in sterile PBS.